Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromoacetaldehyde dimethyl acetal 97+% stabilized

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Bromo-1,1-dimethoxyethane

Stabilization: Stabilised with potassium carbonate

A11324-0B A11324-30 A11324-14 A11324-30 A11324-14 A11324-0B
AAA11324-30EA 178.99 USD InStock
AAA11324-30 CAAAAA11324-14 AAAA11324-14 AAA11324-0B CAAAA11324-0B CAAAA11324-30
Bromoacetaldehyde dimethyl acetal 97+% stabilized
Bromoacetaldehyde dimethyl acetal


Formula: BrH₂CCH(OCH₃)₂
MW: 169.02 g/mol
Boiling Pt: 150 °C
Density: 1.435
Flash Pt: 54 °C(129 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000213
CAS Number: 7252-83-7
EINECS: 230-669-6
UN: 2929
ADR: "6.1",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty