Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Chloro-2-thiophenesulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 5-Chlorothiophene-2-sulphonyl chloride

A11348-09 A11348-06 A11348-03 A11348-09 A11348-06 A11348-03
AAAA11348-09EA 248.99 USD InStock
AAAA11348-09 CAAAAA11348-09 CAAAA11348-03 CAAAA11348-06 AAA11348-03 AAA11348-06
5-Chloro-2-thiophenesulfonyl chloride 97%
5-Chloro-2-thiophenesulphonyl chloride


Formula: C₄H₂Cl₂O₂S₂
MW: 217.1 g/mol
Boiling Pt: 117…120 °C/14 mm
Melting Pt: 28 °C
Density: 1.623
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00051667
CAS Number: 2766-74-7
EINECS: 700-248-6
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty