Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,6-Dichloropyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

A11364-36 A11364-22 A11364-22 A11364-36
AAA11364-22EA 52.13 USD InStock
CAAAA11364-36 AAA11364-22 AAA11364-36 CAAAA11364-22
2,6-Dichloropyridine 98%
2,6-Dichloropyridine


Formula: C₅H₃Cl₂N
MW: 147.99 g/mol
Boiling Pt: 211 °C
Melting Pt: 84…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00006244
CAS Number: 2402-78-0
EINECS: 219-282-3
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty