Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Hexadecyne tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals

A11366-14 A11366-06 A11366-14 A11366-06
AAA11366-14EA 444.99 USD InStock
CAAAA11366-14 CAAAA11366-06 AAA11366-14 AAA11366-06
1-Hexadecyne tech. 90%, Technical Grade
1-Hexadecyne


Formula: C₁₆H₃₀
MW: 222.41 g/mol
Boiling Pt: 146…148 °C/10 mm
Melting Pt: 13…16 °C
Density: 0.790
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015084
CAS Number: 629-74-3
EINECS: 211-106-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty