Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Methylbenzo[b]thiophene-2-carboxaldehyde 98%

Supplier: Thermo Scientific Chemicals

A11375-06 A11375-03 A11375-14 A11375-06 A11375-03
AAA11375-03EA 89.89 USD InStock
AAA11375-03 CAAAAA11375-14 AAA11375-06 CAAAA11375-06 CAAAA11375-03
3-Methylbenzo[b]thiophene-2-carboxaldehyde 98%
3-Methylbenzo[b]thiophene-2-carboxaldehyde


Formula: C₁₀H₈OS
Melting Pt: 88…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00052307
CAS Number: 22053-74-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty