Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloroanisole 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Chloro-4-methoxybenzene

A11377-0B A11377-18 A11377-18 A11377-30 A11377-30 A11377-0B
AAA11377-30EA 237.99 USD InStock
CAAAA11377-0B AAA11377-30 CAAAA11377-30 AAA11377-0B AAA11377-18 CAAAA11377-18
4-Chloroanisole 99%
4-Chloroanisole


Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 198…200 °C
Melting Pt: –18 °C
Density: 1.170
Flash Pt: 78 °C(172 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000616
CAS Number: 623-12-1
EINECS: 210-772-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty