Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloro-3-nitrotoluene 97+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Chloro-4-methyl-2-nitrobenzene

A11383-36 A11383-22 A11383-36 A11383-0E A11383-22 A11383-0E
AAA11383-0EEA 1581.68 USD InStock
AAA11383-0E AAA11383-36 AAA11383-22 CAAAA11383-36 CAAAA11383-0E CAAAA11383-22
4-Chloro-3-nitrotoluene 97+%
4-Chloro-3-nitrotoluene


Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 260…262 °C
Melting Pt: ca 7 °C
Density: 1.295
Flash Pt: 129 °C(264 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007085
CAS Number: 89-60-1
EINECS: 201-922-8
UN: 2433
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty