Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Neocuproine hemihydrate 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2,9-Dimethyl-1,10-phenanthroline hemihydrate

A11398-22 A11398-06 A11398-06 A11398-14 A11398-14 A11398-22
AAA11398-06EA 80.36 USD InStock
CAAAA11398-22 AAA11398-06 CAAAA11398-06 AAA11398-14 AAA11398-22 CAAAA11398-14
Neocuproine hemihydrate 98+%
Neocuproine hemihydrate
Reagent for the colorimetric determination of copper


Formula: C₁₄H₁₂N₂·0,5H₂O
MW: 434,53 g/mol
Melting Pt: 160…164 °C
Storage Temperature: Ambient
MDL Number: MFCD00149306
CAS Number: 34302-69-7
EINECS: 207-601-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty