Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,1,3,3-Tetramethoxypropane 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Malondialdehyde bis(dimethyl acetal) Malondialdehyde tetramethyl acetal

A11418-0B A11418-30 A11418-30 A11418-0I A11418-0B A11418-0I
AAA11418-0IEA 589.99 USD InStock
CAAAA11418-30 AAA11418-0I CAAAA11418-0B AAA11418-30 CAAAA11418-0I AAA11418-0B
1,1,3,3-Tetramethoxypropane 98%
1,1,3,3-Tetramethoxypropane


Formula: C₇H₁₆O₄
MW: 164.2 g/mol
Boiling Pt: 182…183 °C
Density: 0.997
Flash Pt: 54 °C(129 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008488
CAS Number: 102-52-3
EINECS: 203-037-2
UN: 3271
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty