Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-Methyl-2-amino-3-phenylpropanoate hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Synonyms: H-Phe-OMe·HClH-Phe-OMe·HClL-Methyl phenylalanate hydrochloride

A11455-18 A11455-09 A11455-09 A11455-30 A11455-30 A11455-18
AAA11455-18EA 204.99 USD InStock
AAA11455-18 AAA11455-09 CAAAA11455-09 CAAAA11455-18 AAA11455-30 CAAAA11455-30
(S)-Methyl-2-amino-3-phenylpropanoate hydrochloride 98%
(S)-Methyl-2-amino-3-phenylpropanoate hydrochloride


Formula: C₁₀H₁₄ClNO₂
MW: 215.68 g/mol
Melting Pt: 158…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00012489
CAS Number: 7524-50-7
EINECS: 231-383-4
Order Now
Specification Test Results
Optical rotation +37° (c=2 in ethanol)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty