Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Nitrocinnamaldehyde predominantly trans 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-(4-Nitrophenyl)acrylaldehyde

A11467-06 A11467-18 A11467-06 A11467-14 A11467-14 A11467-18
AAAA11467-18EA 803.99 USD InStock
CAAAAA11467-18 AAAA11467-18 CAAAA11467-06 CAAAA11467-14 AAA11467-14 AAA11467-06
4-Nitrocinnamaldehyde predominantly trans 98%
4-Nitrocinnamaldehyde


Formula: C₉H₇NO₃
MW: 177.16 g/mol
Melting Pt: 138…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00007379
CAS Number: 1734-79-8
EINECS: 217-076-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty