Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloroacetic acid 99%

Supplier: Thermo Scientific Chemicals
Danger

A11482-0B A11482-30 A11482-0B A11482-30
AAA11482-30EA 65.6 USD InStock
CAAAA11482-0B AAA11482-30 AAA11482-0B CAAAA11482-30
Chloroacetic acid 99%
Chloroacetic acid


Formula: ClCH₂COOH
MW: 94.5 g/mol
Boiling Pt: 189 °C
Melting Pt: 60…63 °C
Density: 1.404
Flash Pt: 126 °C(258 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002683
CAS Number: 79-11-8
EINECS: 201-178-4
UN: 1751
ADR: "6.1",II
Merck Index: 13,02129
https://us-legacy.vwr.com/stibo/web/std.lang.all/20/30/2302030.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty