Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Iodo-p-toluidine 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Iodo-4-methylaniline 4-Amino-2-iodotoluene

A11485-06 A11485-22 A11485-14 A11485-14 A11485-22 A11485-06
AAA11485-22EA 383.99 USD InStock
AAA11485-22 CAAAA11485-14 CAAAA11485-06 AAA11485-14 CAAAA11485-22 AAA11485-06
3-Iodo-p-toluidine 98%
3-Iodo-p-toluidine


Formula: C₇H₈IN
MW: 233.05 g/mol
Melting Pt: 34…37 °C
Storage Temperature: Ambient
MDL Number: MFCD00047843
CAS Number: 35944-64-0
EINECS: 252-803-2
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty