Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diphenylacetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2,2-Diphenylacetic acid

A11489-22 A11489-22 A11489-36 A11489-36
AAA11489-36EA 225.99 USD InStock
AAA11489-36 AAA11489-22 CAAAA11489-22 CAAAA11489-36
Diphenylacetic acid 99%
Diphenylacetic acid


Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Boiling Pt: 195 °C/25 mm
Melting Pt: 146…148 °C
Density: 1,258 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004251
CAS Number: 117-34-0
EINECS: 204-185-0
Merck Index: 13,03348
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty