Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Nitrobenzaldehyde 98+%

Supplier: Thermo Scientific Chemicals
Warning

A11501-14 A11501-36 A11501-22 A11501-14 A11501-36 A11501-22
AAA11501-14EA 70.57 USD InStock
AAA11501-14 AAA11501-36 AAA11501-22 CAAAA11501-14 CAAAA11501-36 CAAAA11501-22
2-Nitrobenzaldehyde 98+%
2-Nitrobenzaldehyde


Formula: C₇H₅NO₃
MW: 151.12 g/mol
Boiling Pt: 153 °C/23 mm
Melting Pt: 43…46 °C
Density: 1,35 g/cm³ (25 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007132
CAS Number: 552-89-6
EINECS: 209-025-3
Merck Index: 12,06684
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty