Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Pentylphenol 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-Amylphenol

A11514-22 A11514-14 A11514-14 A11514-06 A11514-22 A11514-06
AAA11514-06EA 57.19 USD InStock
AAA11514-06 AAA11514-14 CAAAA11514-14 CAAAA11514-22 AAA11514-22 CAAAA11514-06
4-Pentylphenol 98%
4-Pentylphenol


Formula: C₁₁H₁₆O
MW: 164.25 g/mol
Boiling Pt: 342 °C
Melting Pt: 24 °C
Density: 0.960
Flash Pt: 133 °C(271 °F)
Storage Temperature: Ambient
MDL Number: MFCD00020211
CAS Number: 14938-35-3
EINECS: 239-015-4
UN: 2430
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty