Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-ɑ-D-mannopyranoside 99%

Supplier: Thermo Scientific Chemicals
Synonyms: alpha-Methyl-D-mannosideMethyl alpha-D-Mannoside

A11533-14 A11533-22 A11533-14 A11533-22
AAA11533-22EA 153.92 USD InStock
AAA11533-22 CAAAA11533-14 CAAAA11533-22 AAA11533-14
Methyl-ɑ-D-mannopyranoside 99%
Methyl-alpha-D-mannopyranoside


Formula: C₇H₁₄O₆
MW: 194.18 g/mol
Melting Pt: 193…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00063262
CAS Number: 617-04-9
EINECS: 210-502-3
Order Now
Specification Test Results
Optical rotation +79° (c=4 in water)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty