Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Iodo-2'-deoxyuridine (Idoxuridine) 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-iodopyrimidine-2,4(1H,3H)-dioneIdoxuridine(+)-5-Iodo-2'-deoxyuridine

A11542-06 A11542-03 A11542-06 A11542-03
AAA11542-06EA 238.99 USD InStock
AAA11542-06 AAA11542-03 CAAAA11542-03 CAAAA11542-06
5-Iodo-2'-deoxyuridine (Idoxuridine) 98%
5-Iodo-2'-deoxyuridine (Idoxuridine)


Formula: C₉H₁₁IN₂O₅
MW: 354.10 g/mol
Melting Pt: ca 190 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00134656
CAS Number: 54-42-2
EINECS: 200-207-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty