Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-2-Methylbutyric acid 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (±)-2-Methylbutanoic acid

A11546-AC A11546-0F A11546-AC A11546-AP A11546-AP A11546-0F
AAA11546-0FEA 733.99 USD InStock
CAAAA11546-0F CAAAAA11546-AC AAA11546-0F CAAAA11546-AP AAAA11546-AC AAA11546-AP
(±)-2-Methylbutyric acid 98%
(±)-2-Methylbutyric acid


Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 175…176 °C
Melting Pt: –70 °C
Density: 0.937
Flash Pt: 73 °C(163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002669
CAS Number: 116-53-0
EINECS: 204-145-2
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty