Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

p-Toluamide 98+%

Supplier: Thermo Scientific Chemicals
Synonyms: 4-Methylbenzamide

A11559-22 A11559-22 A11559-14 A11559-14 A11559-36 A11559-36
AAA11559-14EA 34.69 USD InStock
AAA11559-14 AAA11559-36 CAAAA11559-14 CAAAA11559-36 CAAAA11559-22 AAA11559-22
p-Toluamide 98+%
p-Toluamide


Formula: C₈H₉NO
MW: 135.17 g/mol
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 158…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00008002
CAS Number: 619-55-6
EINECS: 210-600-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty