Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(2-Fluorophenyl)acetonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Fluorobenzyl cyanide

A11612-06 A11612-22 A11612-06 A11612-14 A11612-14 A11612-22
AAA11612-22EA 215.99 USD InStock
AAA11612-22 CAAAA11612-06 CAAAA11612-14 AAA11612-14 AAA11612-06 CAAAA11612-22
(2-Fluorophenyl)acetonitrile 97%
(2-Fluorophenyl)acetonitrile


Formula: C₈H₆FN
MW: 135.14 g/mol
Boiling Pt: 235 °C
Density: 1.061
Flash Pt: >100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001897
CAS Number: 326-62-5
EINECS: 206-313-0
UN: 2929
ADR: "6.1",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty