Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pentafluorobenzyl bromide 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: α-Bromo-2,3,4,5,6-pentafluorotoluene 2,3,4,5,6-Pentafluorobenzyl bromide

A11623-22 A11623-14 A11623-22 A11623-06 A11623-14 A11623-06
AAA11623-14EA 205.15 USD InStock
CAAAA11623-06 AAA11623-14 CAAAA11623-14 AAAA11623-22 AAA11623-06 CAAAAA11623-22
Pentafluorobenzyl bromide 98+%
Pentafluorobenzyl bromide


Formula: (C₆F₅)CH₂Br
MW: 260.99 g/mol
Boiling Pt: 174…175 °C
Melting Pt: 18…20 °C
Density: 1.844
Flash Pt: 83 °C(181 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000299
CAS Number: 1765-40-8
EINECS: 217-182-4
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty