Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aesculin sesquihydrate 97%

Supplier: Thermo Scientific Chemicals
Synonyms: 6,7-Dihydroxycumarin-6β-D-glucopyranoside sesquihydrate

A11624-22 A11624-06 A11624-22 A11624-06 A11624-14 A11624-14
AAA11624-22EA 338.99 USD InStock
AAA11624-22 CAAAA11624-06 AAA11624-14 AAA11624-06 CAAAA11624-14 CAAAA11624-22
Aesculin sesquihydrate 97%
Aesculin sesquihydrate
Inhibits chemically induced carcinogenesis in mouse skin and kidney


Formula: C₁₅H₁₆O₉·1,5H₂O
MW: 367.31 g/mol
Melting Pt: 203…205 °C
Storage Temperature: Ambient
MDL Number: MFCD00149492
CAS Number: 66778-17-4
EINECS: 208-517-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty