Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(3,4-Dihydroxyphenyl)propionic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

A11638-22 A11638-06 A11638-14 A11638-06 A11638-14 A11638-22
AAA11638-22EA 648 USD InStock
AAA11638-22 CAAAA11638-22 AAA11638-14 AAA11638-06 CAAAA11638-06 CAAAA11638-14
3-(3,4-Dihydroxyphenyl)propionic acid 98+%
3-(3,4-Dihydroxyphenyl)propionic acid


Formula: C₉H₁₀O₄
MW: 182.18 g/mol
Melting Pt: 136…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00002776
CAS Number: 1078-61-1
EINECS: 214-083-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty