Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bronopol 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Bromo-2-nitropropane-1,3-diol

A11639-22 A11639-36 A11639-22 A11639-36
AAA11639-22EA 99.14 USD InStock
CAAAA11639-36 CAAAA11639-22 AAA11639-22 AAA11639-36
Bronopol 98+%
Bronopol


Formula: HOCH₂BrC(NO₂)CH₂OH
MW: 199.99 g/mol
Melting Pt: ca 122…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00007390
CAS Number: 52-51-7
EINECS: 200-143-0
UN: 3241
ADR: "4.1",III
Merck Index: 13,01432
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty