Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Nitrobenzaldehyde 99%

Supplier: Thermo Scientific Chemicals
Warning

A11655-36 A11655-22 A11655-36 A11655-14 A11655-22
AAA11655-36EA 539.99 USD InStock
AAA11655-36 AAA11655-22 CAAAA11655-22 CAAAA11655-36 CAAAA11655-14
4-Nitrobenzaldehyde 99%
4-Nitrobenzaldehyde


Formula: C₇H₅NO₃
MW: 151.12 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 104…106 °C
Density: 1,546 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007346
CAS Number: 555-16-8
EINECS: 209-084-5
Merck Index: 12,06684
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty