Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Butoxybenzoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-N-butoxybenzoylchloride

A11658-22 A11658-14 A11658-22 A11658-14
AAAA11658-22EA 1298.27 USD InStock
AAAA11658-22 AAA11658-14 CAAAA11658-14 CAAAAA11658-22
4-Butoxybenzoyl chloride 98%
4-Butoxybenzoyl chloride


Formula: C₁₁H₁₃ClO₂
MW: 212.68 g/mol
Boiling Pt: 161…162 °C/9 mm
Density: 1.121
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000689
CAS Number: 33863-86-4
EINECS: 251-703-6
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty