Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Nitro-p-anisidine 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-Amino-3-nitroanisole 4-Methoxy-2-nitroaniline

A11673-0E A11673-0E A11673-22 A11673-36 A11673-36 A11673-22
AAA11673-0EEA 481.99 USD InStock
CAAAA11673-22 AAA11673-0E AAA11673-36 AAA11673-22 CAAAA11673-36 CAAAA11673-0E
2-Nitro-p-anisidine 99%
2-Nitro-p-anisidine


Formula: C₇H₈N₂O₃
MW: 168.15 g/mol
Melting Pt: 126…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00007152
CAS Number: 96-96-8
EINECS: 202-547-2
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty