Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(+)-Malic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: D(+)-Hydroxysuccinic acid(R)-(+)-Hydroxy-1,4-butanedioic acid(R)-2-Hydroxysuccinic acidD(+)-Hydroxy-1,4-butanedioic acidD(+)-Malic acid(R)-(+)-Hydroxysuccinic acid

A11688-14 A11688-06 A11688-06 A11688-14
AAAA11688-14EA 325.99 USD InStock
AAAA11688-14 AAA11688-06 CAAAAA11688-14 CAAAA11688-06
D-(+)-Malic acid 98+%
D(+)-Malic acid


Formula: C₄H₆O₅
MW: 134.09 g/mol
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 100…104 °C
Density: 1.60
Storage Temperature: Ambient
MDL Number: MFCD00004245
CAS Number: 636-61-3
EINECS: 211-262-2
Merck Index: 13,05307
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty