Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Allyl trifluoroacetate 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Trifluoroacetic allyl ester

A11690-09 A11690-18 A11690-30 A11690-30 A11690-09 A11690-18
AAA11690-18EA 141.99 USD InStock
CAAAA11690-30 AAA11690-18 CAAAA11690-09 AAA11690-09 CAAAA11690-18 AAA11690-30
Allyl trifluoroacetate 97%
Allyl trifluoroacetate


Formula: C₅H₅F₃O₂
Boiling Pt: 73…75 °C
Density: 1.183
Flash Pt: -1 °C(30 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013567
CAS Number: 383-67-5
EINECS: 206-853-7
UN: 2924
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty