Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Ethylbenzoyl chloride 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-Ethylbenzene-1-carbonyl chloride

A11698-09 A11698-09 A11698-18 A11698-18
AAA11698-18EA 279.99 USD InStock
AAA11698-18 AAA11698-09 CAAAA11698-18 CAAAA11698-09
4-Ethylbenzoyl chloride 98+%
4-Ethylbenzoyl chloride


Formula: C₉H₉ClO
MW: 168.62 g/mol
Boiling Pt: 235…236 °C
Melting Pt: 42…43 °C
Density: 1.146
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000697
CAS Number: 16331-45-6
EINECS: 240-404-6
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty