Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diethylamine 99+%

Supplier: Thermo Scientific Chemicals
Danger

A11716-AE A11716-AP A11716-0F A11716-AP A11716-0F A11716-AE
AAA11716-0FEA 106.69 USD InStock
AAA11716-0F CAAAAA11716-AE CAAAA11716-0F AAAA11716-AE AAA11716-AP CAAAA11716-AP
Diethylamine 99+%
Diethylamine


Formula: (C₂H₅)₂NH
MW: 73.14 g/mol
Boiling Pt: 54…55 °C
Melting Pt: –50 °C
Density: 0.706
Flash Pt: -28 °C(-18 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009032
CAS Number: 109-89-7
EINECS: 203-716-3
UN: 1154
ADR: "3",II
Merck Index: 13,03138
https://us-legacy.vwr.com/stibo/web/std.lang.all/76/20/2297620.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty