Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Chloro-1,3-dimethyl-1H-pyrazole 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 5-Chloro-1,3-dimethylpyrazole

A11730-14 A11730-06 A11730-06 A11730-14
AAA11730-06EA 111.99 USD InStock
AAA11730-06 CAAAA11730-06 CAAAA11730-14 AAA11730-14
5-Chloro-1,3-dimethyl-1H-pyrazole 98%
5-Chloro-1,3-dimethyl-1H-pyrazole


Formula: C₅H₇ClN₂
Boiling Pt: 153…155 °C
Density: 1,137
Storage Temperature: Ambient
MDL Number: MFCD00051650
CAS Number: 54454-10-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty