Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-δ-Decalactone 97%

Supplier: Thermo Scientific Chemicals
Synonyms: (±)-Decan-5-olide (±)-Tetrahydro-6-pentyl-2H-pyran-2-one

A11756-22 A11756-36 A11756-14 A11756-14 A11756-22 A11756-36
AAA11756-22EA 282.55 USD InStock
AAA11756-22 AAA11756-36 AAA11756-14 CAAAA11756-14 CAAAA11756-36 CAAAA11756-22
(±)-δ-Decalactone 97%
(±)-δ-Decalactone


Formula: C₁₀H₁₈O₂
MW: 170.25 g/mol
Boiling Pt: 117…120 °C/0.02 mm
Melting Pt: –25 °C
Density: 0.971
Flash Pt: 145 °C(293 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006649
CAS Number: 705-86-2
EINECS: 211-889-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty