Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Riboflavine (vitamin B2) 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Lactoflavin Vitamin B2 Vitamine G (-)-Riboflavin

A11764-22 A11764-14 A11764-22 A11764-14
AAA11764-14EA 52.47 USD InStock
AAA11764-14 AAA11764-22 CAAAA11764-22 CAAAA11764-14
Riboflavine (vitamin B2) 98%
Riboflavine (Vitamin B2)
Photoinitiator of polymerization for PAGE gels


Formula: C₁₇H₂₀N₄O₆
MW: 376.37 g/mol
Melting Pt: 290 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005022
CAS Number: 83-88-5
EINECS: 201-507-1
Merck Index: 12,08367
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty