Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-Isonipecotic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (±)-4-Piperidinecarboxylic acid

A11795-22 A11795-14 A11795-14 A11795-22
AAA11795-22EA 120.99 USD InStock
CAAAA11795-22 AAA11795-22 AAA11795-14 CAAAA11795-14
(±)-Isonipecotic acid 98%
(±)-Isonipecotic acid
A GABA-A receptor partial agonist


Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
Melting Pt: >350 °C
Storage Temperature: Ambient
MDL Number: MFCD00006004
CAS Number: 498-94-2
EINECS: 207-872-3
Merck Index: 13,05208
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty