Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nitromethane 98+%

Supplier: Thermo Scientific Chemicals
Warning

A11806-22 A11806-0E A11806-36
AAA11806-0EEA 241.11 USD InStock
AAA11806-0E AAA11806-36 AAA11806-22
Nitromethane 98+%
Nitromethane


Formula: CH₃NO₂
MW: 61.04 g/mol
Boiling Pt: 101 °C
Melting Pt: –29 °C
Density: 1.130
Flash Pt: 35 °C(95 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007400
CAS Number: 75-52-5
EINECS: 200-876-6
UN: 1261
ADR: "3",II
Merck Index: 13,06644
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty