Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Hydroxy-1,4-naphthoquinone 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Hydroxy-p-naphthoquinone2-Hydroxy-1,4-naphoquinone

A11880-18 A11880-30 A11880-18 A11880-30 A11880-09 A11880-09
AAA11880-30EA 352.99 USD InStock
CAAAA11880-18 AAA11880-30 AAA11880-18 AAA11880-09 CAAAA11880-09 CAAAA11880-30
2-Hydroxy-1,4-naphthoquinone 98+%
2-Hydroxy-1,4-naphthoquinone


Formula: C₁₀H₆O₃
MW: 174.16 g/mol
Melting Pt: ca 193 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00001678
CAS Number: 83-72-7
EINECS: 201-496-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty