Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methylesculetin 97%

Supplier: Thermo Scientific Chemicals
Synonyms: 6,7-Dihydroxy-4-methylcoumarin

A11902-22 A11902-22 A11902-14 A11902-14 A11902-06 A11902-06
AAA11902-22EA 344.99 USD InStock
AAA11902-22 CAAAA11902-22 AAA11902-14 AAA11902-06 CAAAA11902-06 CAAAA11902-14
4-Methylesculetin 97%
4-Methylesculetin


Formula: C₁₀H₈O₄
MW: 192.17 g/mol
Melting Pt: ca 275 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006859
CAS Number: 529-84-0
EINECS: 208-470-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty