Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromoform 97% stabilized

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Tribromomethane

Stabilization: Stabilised with ethanol ≤ 3 %

A11904-30 A11904-0I A11904-30 A11904-0B A11904-0B
AAAA11904-30EA 127.99 USD InStock
AAAA11904-30 CAAAAA11904-30 AAA11904-0I AAAA11904-0B CAAAAA11904-0B
Bromoform 97% stabilized
Bromoform


Formula: Br₃CH
MW: 252.73 g/mol
Boiling Pt: 148…150 °C
Melting Pt: 8…9 °C
Density: 2.81
Flash Pt: None
Storage Temperature: Ambient
MDL Number: MFCD00000128
CAS Number: 75-25-2
EINECS: 200-854-6
UN: 2515
ADR: "6.1",III
Merck Index: 13,01404
https://us-legacy.vwr.com/stibo/web/std.lang.all/78/85/717885.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty