Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Arabinose 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Pectin SugarPectinose

A11921-30 A11921-18 A11921-30 A11921-18
AAA11921-18EA 130.47 USD InStock
AAA11921-18 CAAAA11921-30 CAAAA11921-18 AAA11921-30
L(+)-Arabinose 99%
L(+)-Arabinose


Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Melting Pt: 155…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00067709
CAS Number: 5328-37-0
EINECS: 201-767-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/24/52/2302452.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty