Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Thiophenecarbonitrile 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Cyanothiophene 2-Thienyl cyanide Thiophene-2-carbonitrile

A11930-22 A11930-14 A11930-22 A11930-36 A11930-36 A11930-14
AAA11930-14EA 139.99 USD InStock
AAA11930-14 AAA11930-22 CAAAA11930-22 CAAAA11930-14 AAAA11930-36 CAAAAA11930-36
2-Thiophenecarbonitrile 98%
2-Thiophenecarbonitrile


Formula: C₅H₃NS
MW: 109.15 g/mol
Boiling Pt: 191…193 °C
Density: 1.171
Flash Pt: 81 °C(178 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005416
CAS Number: 1003-31-2
EINECS: 213-706-0
UN: 1992
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty