Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3-Diaminopropane 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1,3-Propanediamine Trimethylenediamine

A11932-AE A11932-AE A11932-AP A11932-0F A11932-AP A11932-0F
AAA11932-0FEA 228.99 USD InStock
CAAAAA11932-AE AAA11932-0F CAAAA11932-AP AAA11932-AP CAAAA11932-0F AAAA11932-AE
1,3-Diaminopropane 98%
1,3-Diaminopropane


Formula: C₃H₁₀N₂
MW: 74.13 g/mol
Boiling Pt: 134…136 °C
Melting Pt: –12 °C
Density: 0.888
Flash Pt: 48 °C(118 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008228
CAS Number: 109-76-2
EINECS: 203-702-7
UN: 2734
ADR: "8",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty