Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-1,2-difluorobenzene 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Bromo-3,4-difluorobenzene

A11934-06 A11934-22 A11934-14 A11934-22 A11934-14 A11934-06
AAA11934-06EA 44.39 USD InStock
CAAAA11934-22 AAA11934-06 AAA11934-22 CAAAA11934-14 AAA11934-14 CAAAA11934-06
4-Bromo-1,2-difluorobenzene 98+%
4-Bromo-1,2-difluorobenzene


Formula: C₆H₃BrF₂
MW: 192.99 g/mol
Boiling Pt: 150…151 °C
Melting Pt: –4 °C
Density: 1.707
Flash Pt: 33 °C(91 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000304
CAS Number: 348-61-8
EINECS: 206-481-5
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty