Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluorophenol 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Fluoro-4-hydroxybenzene

A11945-36 A11945-22 A11945-22 A11945-36 A11945-14 A11945-14
AAA11945-22EA 116.99 USD InStock
CAAAA11945-22 CAAAA11945-36 AAA11945-22 CAAAA11945-14 AAA11945-36 AAA11945-14
4-Fluorophenol 99%
4-Fluorophenol


Formula: C₆H₅FO
MW: 112.1 g/mol
Boiling Pt: 185…188 °C
Melting Pt: 44…50 °C
Density: 1.22
Flash Pt: 68 °C(154 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002316
CAS Number: 371-41-5
EINECS: 206-736-0
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty