Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-Phenylsuccinic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (±)-Phenyl-1,4-butanedioic acid

A11971-14 A11971-36 A11971-36 A11971-22 A11971-14 A11971-22
AAA11971-36EA 383.99 USD InStock
CAAAA11971-22 CAAAA11971-14 AAA11971-36 CAAAA11971-36 AAA11971-14 AAA11971-22
(±)-Phenylsuccinic acid 98%
(±)-Phenylsuccinic acid


Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
Boiling Pt: 368 °C (1013 hPa)
Melting Pt: 167…169 °C
Storage Temperature: Ambient
MDL Number: MFCD00004256
CAS Number: 635-51-8
EINECS: 211-238-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty