Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Deoxy-D(-)-ribose 99%

Supplier: Thermo Scientific Chemicals
Synonyms: 2-Deoxy-D(-)-arabinose 2-Deoxy-D(-)-erythropentose Thyminose

A11990-14 A11990-03 A11990-06 A11990-14 A11990-06 A11990-03
AAA11990-14EA 220.99 USD InStock
AAA11990-14 AAA11990-03 CAAAA11990-06 AAA11990-06 CAAAA11990-03 CAAAA11990-14
2-Deoxy-D(-)-ribose 99%
2-Deoxy-D(-)-ribose


Formula: C₅H₁₀O₄
MW: 134.13 g/mol
Boiling Pt: 292 °C (1013 hPa)
Melting Pt: 76…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00135904
CAS Number: 533-67-5
EINECS: 208-573-0
Merck Index: 13,02925
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty