Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chlorophenyl isothiocyanate 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1-Chloro-3-isothiocyanatobenzene

A11999-22 A11999-06 A11999-14 A11999-06 A11999-22 A11999-14
AAA11999-22EA 488.53 USD InStock
AAA11999-22 CAAAA11999-14 CAAAA11999-06 AAA11999-14 CAAAA11999-22 AAA11999-06
3-Chlorophenyl isothiocyanate 97%
3-Chlorophenyl isothiocyanate


Formula: C₇H₄ClNS
MW: 169.63 g/mol
Boiling Pt: 249…250 °C
Melting Pt: 45 °C
Density: 1.281
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004805
CAS Number: 2392-68-9
EINECS: 000-000-0
UN: 2810
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty