Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Octadecanol 97%

Supplier: Thermo Scientific Chemicals
Synonyms: 1-Octadecyl alcohol Heptadecyl carbinol Stearyl alcohol

A12020-30 A12020-36 A12020-36 A12020-30 A12020-0E A12020-0E
AAA12020-36EA 49.11 USD InStock
CAAAA12020-0E CAAAAA12020-30 CAAAA12020-36 AAA12020-36 AAAA12020-30 AAA12020-0E
1-Octadecanol 97%
1-Octadecanol


Formula: H₃C(CH₂)₁₆CH₂OH
MW: 270.5 g/mol
Boiling Pt: 210 °C/15 mm
Melting Pt: 56…60 °C
Density: 0.805…0.815 g/cm³ (60 °C)
Flash Pt: 174 °C(345 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002823
CAS Number: 112-92-5
EINECS: 204-017-6
Merck Index: 12,08960
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty