Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formaldehyde dimethyl acetal 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Dimethoxymethane Methylal Formal

A12055-0F A12055-AE A12055-AP A12055-0F A12055-AP A12055-AE
AAA12055-0FEA 107.51 USD InStock
CAAAAA12055-AE AAA12055-0F CAAAA12055-AP AAA12055-AP CAAAA12055-0F AAAA12055-AE
Formaldehyde dimethyl acetal 98%
Formaldehyde dimethyl acetal


Formula: CH₂(OCH₃)₂
MW: 76.1 g/mol
Boiling Pt: 41…42 °C
Melting Pt: –105 °C
Density: 0.863
Flash Pt: -17 °C(1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008495
CAS Number: 109-87-5
EINECS: 203-714-2
UN: 1234
ADR: "3",II
Merck Index: 12,06093
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/92/730692.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty